Assay Details
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567034.2 | XP_005567091.1 | 7 - 8 | 714 | 86 | |
XM_005567035.2 | XP_005567092.1 | 8 - 9 | 1198 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
