Assay Details
| Gene Symbol: CD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589885.2 | XP_005589942.1 | 3 - 4 | 412 | 99 | |
| XM_005589886.2 | XP_005589943.1 | 3 - 4 | 402 | 99 | ||
More Information
Set Membership: |
|

CD37
Genomic Map