Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287722.1 | NP_001274651.1 | 4 - 5 | 371 | 131 | |
| GenBank mRNA | AB093680.1 | - | 4 - 5 | 371 | 131 | |
| AB170121.1 | - | 4 - 5 | 371 | 131 | ||
More Information
Set Membership: |
|

RPL30
Genomic Map