Assay Details
| Gene Symbol: ATP5MG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579802.1 | XP_005579859.1 | 1 - 2 | 334 | 141 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005579802.1 transcripts.
Set Membership: |
|

ATP5MG
Genomic Map