Assay Details
| Gene Symbol: GSPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591279.2 | XP_005591336.1 | 7 - 8 | 1079 | 109 | |
| XM_005591280.2 | XP_005591337.1 | 7 - 8 | 1076 | 109 | ||
More Information
Set Membership: |
|

GSPT1
Genomic Map