Assay Details
| Gene Symbol: LOC102142492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554238.2 | XP_005554295.1 | 3 - 4 | 434 | 76 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  3' Most  Recommended Assay  Probe spans exons | 
Panther Classification:
 Back To Top
 Back To Top
 LOC102142492
LOC102142492 Genomic Map
Genomic Map Back To Top
 Back To Top