Assay Details
| Gene Symbol: UBE2L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577852.2 | XP_005577909.1 | 3 - 4 | 497 | 66 | |
| XM_015434702.1 | XP_015290188.1 | 3 - 4 | 768 | 66 | ||
More Information
Set Membership: |
|

UBE2L6
Genomic Map