Assay Details
| Gene Symbol: ISCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582070.2 | XP_005582127.1 | 4 | 695 | 131 | |
| GenBank mRNA | AB169593.1 | - | 4 | 498 | 131 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ISCA1| Gene Symbol: ISCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582070.2 | XP_005582127.1 | 4 | 695 | 131 | |
| GenBank mRNA | AB169593.1 | - | 4 | 498 | 131 | |
Set Membership: |
|