Assay Details
Gene Symbol: TMEM35B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544012.2 | XP_005544069.1 | 3 | 1189 | 147 | |
XM_005544013.2 | XP_005544070.1 | 3 | 854 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
