Assay Details
Gene Symbol: TAS2R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550985.2 | XP_005551042.1 | 1 | 2364 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005550985.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
