Assay Details
| Gene Symbol: LRRC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580702.2 | XP_005580759.1 | 3 - 4 | 2412 | 117 | |
| XM_005580703.2 | XP_005580760.1 | 3 - 4 | 2627 | 117 | ||
| XM_005580704.2 | XP_005580761.1 | 2 - 3 | 3779 | 117 | ||
| XM_005580705.2 | XP_005580762.1 | 2 - 3 | 2305 | 117 | ||
More Information
Set Membership: |
|

LRRC8A
Genomic Map