Assay Details
| Gene Symbol: SLC26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015450005.1 | XP_015305491.1 | 2 - 3 | 1507 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC26A1| Gene Symbol: SLC26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015450005.1 | XP_015305491.1 | 2 - 3 | 1507 | 90 | |
Set Membership: |
|