Assay Details
| Gene Symbol: SYNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544085.2 | XP_005544142.1 | 2 - 3 | 1345 | 143 | |
| XM_005544086.2 | XP_005544143.1 | 2 - 3 | 1345 | 143 | ||
| XM_005544087.2 | XP_005544144.1 | 2 - 3 | 1345 | 143 | ||
| XM_015439002.1 | XP_015294488.1 | 2 - 3 | 1293 | 143 | ||
More Information
Set Membership: |
|

SYNC
Genomic Map