Assay Details
Gene Symbol: TMEM121B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568060.2 | XP_005568117.1 | 2 | 1205 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM121B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568060.2 | XP_005568117.1 | 2 | 1205 | 56 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |