Assay Details
Gene Symbol: USP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015444353.1 | XP_015299839.1 | 1 | 2090 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
