Assay Details
Gene Symbol: LRRC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545613.2 | XP_005545670.1 | 1 - 2 | 426 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005545613.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
