Assay Details
| Gene Symbol: SLC32A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285396.1 | NP_001272325.1 | 1 - 2 | 829 | 108 | |
| GenBank mRNA | AB062931.1 | - | 1 - 2 | 829 | 108 | |
More Information
Set Membership: |
|

SLC32A1
Genomic Map