Assay Details
Gene Symbol: SPSB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545941.2 | XP_005545998.1 | 1 - 2 | 678 | 81 | |
XM_005545942.2 | XP_005545999.1 | 1 - 2 | 686 | 81 | ||
XM_005545943.2 | XP_005546000.1 | 1 - 2 | 687 | 81 | ||
XM_015445149.1 | XP_015300635.1 | 1 - 2 | 687 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005545941.2, XM_005545942.2, XM_005545943.2, XM_015445149.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
