Assay Details
Gene Symbol: HEXIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015438634.1 | XP_015294120.1 | 2 - 3 | 1947 | 122 | |
XM_015438637.1 | XP_015294123.1 | 3 - 4 | 1491 | 122 | ||
XM_015438639.1 | XP_015294125.1 | 3 - 4 | 1399 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015438634.1, XM_015438637.1, XM_015438639.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
