Assay Details
| Gene Symbol: C16H17orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583481.2 | XP_005583538.1 | 2 - 3 | 1178 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16H17orf50| Gene Symbol: C16H17orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583481.2 | XP_005583538.1 | 2 - 3 | 1178 | 122 | |
Set Membership: |
|