Assay Details
Gene Symbol: SMIM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555829.2 | XP_005555886.1 | 1 - 2 | 224 | 141 | |
XM_005555835.2 | XP_005555892.1 | 1 - 2 | 224 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005555829.2, XM_005555835.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
