Assay Details
| Gene Symbol: COMMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575774.2 | XP_005575831.1 | 3 | 546 | 111 | |
| XM_015433608.1 | XP_015289094.1 | 4 | 562 | 111 | ||
More Information
Set Membership: |
|

COMMD1
Genomic Map