Assay Details
| Gene Symbol: C1H1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542359.2 | XP_005542416.1 | 4 - 5 | 423 | 107 | |
| XM_015431241.1 | XP_015286727.1 | 3 - 4 | 360 | 107 | ||
More Information
Set Membership: |
|

C1H1orf162
Genomic Map