Assay Details
Gene Symbol: LOC107128329 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442241.1 | XP_015297727.1 | 2 - 3 | 146 | 125 |
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442238.1 | XP_015297724.1 | 2 - 3 | 131 | 125 | |
GenBank mRNA | AB066558.1 | - | 2 - 3 | 131 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015442238.1, XM_015442241.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
