Assay Details
| Gene Symbol: HMBS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579888.2 | XP_005579945.1 | 2 - 3 | 255 | 52 | |
| XM_005579890.2 | XP_005579947.1 | 2 - 3 | 949 | 52 | ||
| XM_015435824.1 | XP_015291310.1 | 2 - 3 | 348 | 52 | ||
More Information
Set Membership: |
|

HMBS
Genomic Map