Assay Details
Gene Symbol: GUCA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449404.1 | XP_015304890.1 | 2 - 3 | 2047 | 102 | |
XM_015449405.1 | XP_015304891.1 | 3 - 4 | 1951 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
