Assay Details
| Gene Symbol: PTER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564715.2 | XP_005564772.1 | 5 - 6 | 1127 | 97 | |
| XM_005564716.2 | XP_005564773.1 | 5 - 6 | 1111 | 97 | ||
| XM_005564717.2 | XP_005564774.1 | 5 - 6 | 3633 | 97 | ||
| XM_005564719.2 | XP_005564776.1 | 4 - 5 | 908 | 97 | ||
| XM_015455602.1 | XP_015311088.1 | 4 - 5 | 1149 | 97 | ||
| XM_015455603.1 | XP_015311089.1 | 4 - 5 | 965 | 97 | ||
| XM_015455604.1 | XP_015311090.1 | 5 - 6 | 1022 | 97 | ||
| XM_015455607.1 | XP_015311093.1 | 5 - 6 | 3452 | 97 | ||
| XM_015455608.1 | XP_015311094.1 | 4 - 5 | 724 | 97 | ||
More Information
Set Membership: |
|

PTER
Genomic Map