Assay Details
Gene Symbol: TM6SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588534.2 | XP_005588591.1 | 2 - 3 | 282 | 117 | |
XM_005588535.2 | XP_005588592.1 | 3 - 4 | 340 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005588535.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
