Assay Details
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563059.2 | XP_005563116.1 | 5 - 6 | 3659 | 148 | |
XM_005563060.2 | XP_005563117.1 | 4 - 5 | 1788 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
