Assay Details
Gene Symbol: ISX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567474.2 | XP_005567531.1 | 4 - 5 | 1323 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
