Assay Details
Gene Symbol: ZNF311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553708.2 | XP_005553765.1 | 5 - 6 | 725 | 138 | |
XM_015449695.1 | XP_015305181.1 | 5 - 6 | 939 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
