Assay Details
Gene Symbol: XIRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546707.2 | XP_005546764.1 | 2 | 3946 | 136 | |
XM_005546708.2 | XP_005546765.1 | 2 | 4022 | 136 | ||
XM_005546709.2 | XP_005546766.1 | 2 | 3943 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
