Assay Details
| Gene Symbol: B3GNT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589355.2 | XP_005589412.1 | 1 - 2 | 1686 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005589355.2 transcripts.
Set Membership: |
|

B3GNT8
Genomic Map