Assay Details
| Gene Symbol: C1H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015432877.1 | XP_015288363.1 | 3 - 4 | 546 | 104 | |
| XM_015432917.1 | XP_015288403.1 | 3 - 4 | 573 | 104 | ||
| XM_015432967.1 | XP_015288453.1 | 3 - 4 | 734 | 104 | ||
More Information
Set Membership: |
|

C1H1orf100
Genomic Map