Assay Details
Gene Symbol: ODF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015457015.1 | XP_015312501.1 | 4 - 5 | 1758 | 156 | |
XM_015457016.1 | XP_015312502.1 | 3 - 4 | 1004 | 156 | ||
XM_015457017.1 | XP_015312503.1 | 5 - 6 | 1626 | 156 | ||
XM_015457018.1 | XP_015312504.1 | 3 - 4 | 1554 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
