Assay Details
| Gene Symbol: POLR2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559644.2 | XP_005559701.1 | 3 - 4 | 1285 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR2M| Gene Symbol: POLR2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559644.2 | XP_005559701.1 | 3 - 4 | 1285 | 123 | |
Set Membership: |
|