Assay Details
Gene Symbol: MAJIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577331.2 | XP_005577388.1 | 9 - 10 | 754 | 122 | |
XM_005577332.2 | XP_005577389.1 | 9 - 10 | 702 | 122 | ||
XM_005577333.2 | XP_005577390.1 | 9 - 10 | 666 | 122 | ||
XM_005577337.2 | XP_005577394.1 | 8 - 9 | 534 | 122 | ||
XM_015434497.1 | XP_015289983.1 | 8 - 9 | 690 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
