Assay Details
Gene Symbol: SYNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441579.1 | XP_015297065.1 | 2 - 3 | 610 | 142 | |
XM_015441580.1 | XP_015297066.1 | 2 - 3 | 610 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
