Assay Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586988.2 | XP_005587045.1 | 1 - 2 | 181 | 68 | |
| GenBank mRNA | AB169541.1 | - | 1 - 2 | 86 | 68 | |
| AB170764.1 | - | 1 - 2 | 84 | 68 | ||
| AB173939.1 | - | 1 - 2 | 49 | 68 | ||
More Information
Set Membership: |
|

AQP4
Genomic Map