Assay Details
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553014.2 | XP_005553071.1 | 4 - 5 | 901 | 104 | |
XM_005553017.2 | XP_005553074.1 | 4 - 5 | 1010 | 104 | ||
XM_005553018.2 | XP_005553075.1 | 3 - 4 | 794 | 104 | ||
GenBank mRNA | AB171673.1 | - | 4 - 5 | 550 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
