Assay Details
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005542657.2 | XP_005542714.1 | 3 - 4 | 650 | 100 | |
XM_005542658.2 | XP_005542715.1 | 3 - 4 | 321 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
