Assay Details
| Gene Symbol: KLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285202.1 | NP_001272131.1 | 3 - 4 | 527 | 154 | |
| GenBank mRNA | L10039.1 | - | 3 - 4 | 527 | 154 | |
More Information
Set Membership: |
|

KLK1
Genomic Map