Assay Details
Gene Symbol: PTPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569339.2 | XP_005569396.1 | 9 - 10 | 1487 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
