Assay Details
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583543.2 | XP_005583600.1 | 2 - 3 | 1205 | 103 | |
XM_005583544.2 | XP_005583601.1 | 2 - 3 | 1155 | 103 | ||
XM_005583545.2 | XP_005583602.1 | 2 - 3 | 1205 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
