Assay Details
| Gene Symbol: ZNF76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553231.2 | XP_005553288.1 | 11 - 12 | 1716 | 144 | |
| XM_005553232.2 | XP_005553289.1 | 11 - 12 | 1679 | 144 | ||
| XM_015449530.1 | XP_015305016.1 | 12 - 13 | 1648 | 144 | ||
| XM_015449531.1 | XP_015305017.1 | 12 - 13 | 1781 | 144 | ||
| XM_015449532.1 | XP_015305018.1 | 12 - 13 | 1630 | 144 | ||
| XM_015449533.1 | XP_015305019.1 | 12 - 13 | 1763 | 144 | ||
| XM_015449534.1 | XP_015305020.1 | 11 - 12 | 1564 | 144 | ||
More Information
Set Membership: |
|

ZNF76
Genomic Map