Assay Details
| Gene Symbol: EIF3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284779.1 | NP_001271708.1 | 5 - 6 | 771 | 100 | |
| GenBank mRNA | AB169626.1 | - | 5 - 6 | 771 | 100 | |
More Information
Set Membership: |
|

EIF3F
Genomic Map