Assay Details
| Gene Symbol: CCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564118.2 | XP_005564175.1 | 2 - 3 | 560 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCN4| Gene Symbol: CCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564118.2 | XP_005564175.1 | 2 - 3 | 560 | 68 | |
Set Membership: |
|