Assay Details
| Gene Symbol: UBE2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590626.1 | XP_005590683.1 | 5 - 6 | 463 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2M| Gene Symbol: UBE2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590626.1 | XP_005590683.1 | 5 - 6 | 463 | 116 | |
Set Membership: |
|