Assay Details
| Gene Symbol: SLC7A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442930.1 | XP_015298416.1 | 9 - 10 | 1129 | 177 | |
| XM_015442931.1 | XP_015298417.1 | 9 - 10 | 1299 | 177 | ||
More Information
Set Membership: |
|

SLC7A6
Genomic Map