Assay Details
| Gene Symbol: GNA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581975.2 | XP_005582032.1 | 3 - 4 | 1895 | 116 | |
| XM_005581976.2 | XP_005582033.1 | 2 - 3 | 394 | 116 | ||
More Information
Set Membership: |
|

GNA14
Genomic Map